Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUF2 Monoclonal Antibody (GT644)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531533
Description
Keep as concentrated solution. Positive Control: HeLa, HeLa NE. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
NUF2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9BZD4 | |
NUF2 | |
Recombinant protein corresponding to human NUF2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot | |
GT644 | |
Unconjugated | |
NUF2 | |
2410003C07Rik; AU044112; C85691; cancer/testis antigen 106; Cdca1; cell division cycle associated 1; cell division cycle-associated protein 1; CT106; hNuf2; hNuf2R; hsNuf2; kinetochore protein Nuf2; Nuf2; NUF2, NDC80 kinetochore complex component; NUF2, NDC80 kinetochore complex component, homolog; NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae); NUF2R | |
Mouse | |
Protein A | |
RUO | |
83540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction