Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107201 Supplier Invitrogen™ Supplier No. PA5107201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Rabbit: 93%; Rat: 93%.

This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene NUF2
Gene Accession No. Q9BZD4
Gene Alias 2410003C07Rik; AU044112; C85691; cancer/testis antigen 106; Cdca1; cell division cycle associated 1; cell division cycle-associated protein 1; CT106; hNuf2; hNuf2R; hsNuf2; kinetochore protein Nuf2; Nuf2; NUF2, NDC80 kinetochore complex component; NUF2, NDC80 kinetochore complex component, homolog; NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae); NUF2R
Gene Symbols NUF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of human NUF2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.