Learn More
Invitrogen™ NUF2 Polyclonal Antibody

Description
Recommended positive controls: HeLa, HeLa nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | NUF2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | NUF2 |
| Gene Accession No. | Q9BZD4 |
| Gene Alias | 2410003C07Rik; AU044112; C85691; cancer/testis antigen 106; Cdca1; cell division cycle associated 1; cell division cycle-associated protein 1; CT106; hNuf2; hNuf2R; hsNuf2; kinetochore protein Nuf2; Nuf2; NUF2, NDC80 kinetochore complex component; NUF2, NDC80 kinetochore complex component, homolog; NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae); NUF2R |
| Gene Symbols | NUF2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.