Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUF2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567418
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7418 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7418 Supplier Bethyl Laboratories, Inc Supplier No. A304318AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Nuf2 (also known as CDCA1 or cell division cycle associated 1) is important for the formation of stable kinetochore-microtubule attachments, chromosome alignment in mitosis, and mitotic progression. Nuf2 is a spindle pole body-associated protein required for nuclear division, spindle pole body separation, and spindle elongation. Nuf2 is part of the Ndc80 complex, which is also comprised of Ndc80 (Hec1), Spc24, and Spc25. The Ndc80 complex is essential for metaphase chromosome alignment and anaphase chromosome segregation.

Specifications

Antigen NUF2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NUF2
Gene Accession No. Q9BZD4
Gene Alias CDCA1, CT106, NUF2R, cancer/testis antigen 106, cell division cycle associated 1, cell division cycle-associated protein 1, hNuf2, hNuf2R, hsNuf2, kinetochore protein Nuf2
Gene Symbols NUF2
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83540
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.