Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuf2 Rabbit anti-Human, Clone: 5J0T9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Nuf2 |
---|---|
Clone | 5J0T9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Nuf2 Monoclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western BlotSpecifications
Nuf2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
83540 | |
IgG | |
Affinity purified |
5J0T9 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Nuf2 (Q9BZD4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title