Learn More
Invitrogen™ NUF2 Recombinant Rabbit Monoclonal Antibody (5J0T9)

Description
Positive Samples: HeLa, 293T, MCF7 Immunogen sequence: METLSFPRYN VAEIVIHIRN KILTGADGKN LTKNDLYPNP KPEVLHMIYM RALQIVYGIR LEHFYMMPVN SEVMYPHLME GFLPFSNLVT HLDSFLPICR.
Specifications
Specifications
| Antigen | NUF2 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 5J0T9 |
| Concentration | 0.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NUF2 |
| Gene Accession No. | Q9BZD4 |
| Gene Alias | 2410003C07Rik; AU044112; C85691; cancer/testis antigen 106; Cdca1; cell division cycle associated 1; cell division cycle-associated protein 1; CT106; hNuf2; hNuf2R; hsNuf2; kinetochore protein Nuf2; Nuf2; NUF2, NDC80 kinetochore complex component; NUF2, NDC80 kinetochore complex component, homolog; NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae); NUF2R |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.