Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUFIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556308 Supplier Invitrogen™ Supplier No. PA556308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYYPRKYDAK FTDFSLPPSR KQKKKKRKEP VFHFFCDTCD RGFKNQEKYD KHMSEHTKCP ELDCSFTAHE KIVQFHWRNM Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

This gene encodes a nuclear RNA binding protein that contains a C2H2 zinc finger motif and a nuclear localization signal. This protein is associated with the nuclear matrix in perichromatin fibrils and, in neurons, localizes to the cytoplasm in association with endoplasmic reticulum ribosomes. This protein interacts with the fragile X mental retardation protein (FMRP), the tumor suppressor protein BRCA1, upregulates RNA polymerase II transcription, and is involved in box C/D snoRNP biogenesis. A pseudogene of this gene resides on chromosome 6q12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUFIP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nufip1
Gene Accession No. Q9UHK0
Gene Alias bA540M5.1; nuclear FMRP interacting protein; nuclear FMRP-interacting protein 1; nuclear fragile X mental retardation protein interacting protein 1; nuclear fragile X mental retardation-interacting protein 1; Nufip; NUFIP1; NUFIP1, FMR1 interacting protein 1
Gene Symbols Nufip1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUFIP1. Recombinant protein control fragment (Product #RP-95497).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26747
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.