Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUFIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566922 Supplier Invitrogen™ Supplier No. PA566922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSGYITNGYMG KGADNDGSGS ESGYTTPKKR KARRNSAKGC ENLNIVQDKI MQQETSVPTL KQGLETFKPD YSEQKGNRVD GSKPIWKYET G Highest antigen sequence identity to the following orthologs - mouse 92%, rat 93%.

NUFIP2 binds RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUFIP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nufip2
Gene Accession No. Q7Z417
Gene Alias 1110001M19Rik; 182-FIP; 82 kDa FMRP-interacting protein; 82-FIP; 82-kD FMRP Interacting Protein; 9530056D24Rik; AI256692; AI461731; Cell proliferation-inducing gene 1 protein; FIP-82; FMRP-interacting protein 2; Fragile X Mental Retardation Protein; im:7136583; KIAA1321; LOW QUALITY PROTEIN: nuclear fragile X mental retardation-interacting protein 2; mKIAA1321; nuclear FMR1 interacting protein 2; nuclear FMR1 interacting protein 2; nuclear fragile X mental retardation-interacting protein 2; nuclear fragile X mental retardation protein interacting protein 2; nuclear fragile X mental retardation-interacting protein 2; NUFIP2; NUFIP2, FMR1 interacting protein 2; PIG1; proliferation-inducing gene 1
Gene Symbols Nufip2
Host Species Rabbit
Immunogen Recombinant Human NUFIP2. Recombinant protein control fragment (Product #RP-107575).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.