Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUGGC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549521
Description
NUGGC Polyclonal Antibody for Western Blot
Nuclear GTPase found in germinal center B-cells, where it may inhibit function of the activation-induced cytidine deaminase AICDA (PubMed:19734146). Reduces somatic hypermutation in B-cells which may enhance genome stability. [UniProt].
Specifications
| NUGGC | |
| Polyclonal | |
| Unconjugated | |
| NUGGC | |
| C8orf80; GTPase SLIP-GC; HMFN0672; Nuclear GTPase SLIP-GC; nuclear GTPase, germinal center associated; NUGGC; SLIP-GC; speckled-like pattern in the germinal center | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 389643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q68CJ6 | |
| NUGGC | |
| KLH conjugated synthetic peptide between 279-311 amino acids from the Central region of human NUGGC. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction