Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUGGC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598486 Supplier Invitrogen™ Supplier No. PA598486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUGGC Polyclonal Antibody for Western Blot, ELISA

Nuclear GTPase found in germinal center B-cells, where it may inhibit function of the activation-induced cytidine deaminase AICDA (PubMed:19734146). Reduces somatic hypermutation in B-cells which may enhance genome stability. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUGGC
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NUGGC
Gene Accession No. Q68CJ6
Gene Alias C8orf80; GTPase SLIP-GC; HMFN0672; Nuclear GTPase SLIP-GC; nuclear GTPase, germinal center associated; NUGGC; SLIP-GC; speckled-like pattern in the germinal center
Gene Symbols NUGGC
Host Species Rabbit
Immunogen Recombinant Human Nuclear GTPase SLIP-GC protein (1-228AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389643
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.