Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NULP1 Antibody (PCRP-TCF25-1A11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308294B
Description
NULP1 Monoclonal antibody specifically detects NULP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein ArraySpecifications
NULP1 | |
Monoclonal | |
Biotin | |
hKIAA1049, KIAA1049Hulp1, Nuclear localized protein 1, Nulp1, NULP1PRO2620, TCF-25, transcription factor 25, transcription factor 25 (basic helix-loop-helix) | |
Recombinant full-length human NULP1 protein (Uniprot: Q9BQ70) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Protein Array | |
PCRP-TCF25-1A11 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
22980 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction