Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NULP1 Antibody (PCRP-TCF25-1A11), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308294JF646
Description
NULP1 Monoclonal antibody specifically detects NULP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein ArraySpecifications
| NULP1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| hKIAA1049, KIAA1049Hulp1, Nuclear localized protein 1, Nulp1, NULP1PRO2620, TCF-25, transcription factor 25, transcription factor 25 (basic helix-loop-helix) | |
| Recombinant full-length human NULP1 protein (Uniprot: Q9BQ70) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Protein Array | |
| PCRP-TCF25-1A11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 22980 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction