Learn More
Nulp1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303087AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
TCF25 is a member of the basic helix-loop-helix (bHLH) family of transcription factors that are important in embryonic development (Steen and Lindholm, 2008 [PubMed 18068114]).Specifications
Nulp1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8R3L2, Q9BQ70 | |
TCF25 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TCF25 | |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049, NULP1, TCF-25, nuclear localized protein 1, transcription factor 25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22980, 66855 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.