Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NuMA Antibody (SR1359), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | NuMA |
---|---|
Clone | SR1359 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
NuMA Monoclonal antibody specifically detects NuMA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
NuMA | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein | |
A synthesized peptide derived from human NuMA (Uniprot #: Q14980) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1359 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4926 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title