Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NuMA1 Mouse anti-Human, Mouse, MilliporeSigma™

Catalog No. MABE180725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1807-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1807-25 Supplier MilliporeSigma Supplier No. MABE180725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NuMA1, clone AD6-1 Antibody is a highly specific mouse monoclonal antibody, that targets NUMA1 and has been tested in Western Blotting and Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NuMA1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols NUMA1,NMP22,NUMA
Host Species Mouse
Immunogen A recombinant fragment corresponding to 181 amino acids from the C-terminal region of human Nuclear mitotic apparatus protein 1 (NuMA1).
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone AD6-1 detects NuMA1 in human and mouse cells. It targets an epitope within 181 amino acids from the C-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.