Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Numb Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500178UV
Description
Numb Polyclonal specifically detects Numb in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Numb | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
Rabbit | |
Affinity Purified | |
Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
8650.0 | |
Human, Mouse, Chicken | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
DyLight 350 | |
50 mM sodium borate with 0.05% sodium azide | |
NUMB | |
A synthetic peptide made to a C-terminal region of mouse NUMB (between residues 600-653). [UniProtNo. Q9QZS3] | |
0.1 mL | |
Primary | |
This is specific for all four isoforms of the NUMB protein. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction