Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Numb Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500178F
Description
Numb Polyclonal specifically detects Numb in Human, Mouse, Chicken samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Numb | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for all four isoforms of the NUMB protein. | |
Store at 4C in the dark. |
Immunocytochemistry, Immunofluorescence | |
FITC | |
PBS with 0.05% Sodium Azide | |
NUMB | |
A synthetic peptide made to a C-terminal region of mouse NUMB (between residues 600-653). [UniProt# Q9QZS3] | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
8650 | |
Human, Mouse, Chicken | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction