Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUMB Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
SDP

Catalog No. 502542219
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2219 30 μL
50-167-2659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2219 Supplier OriGene Supplier No. TA501518S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUMB Monoclonal antibody specifically detects NUMB in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen NUMB
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FSHB
Gene Accession No. P01225
Gene Alias c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171
Host Species Mouse
Immunogen Full length human recombinant protein of human FSHB produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.