Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUMBL Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10088136AF405
Description
NUMBL Polyclonal antibody specifically detects NUMBL in Human samples. It is validated for Western BlotSpecifications
NUMBL | |
Polyclonal | |
Western Blot | |
CAG3A, CTG3a, numb (Drosophila) homolog-like, numb homolog (Drosophila)-like, numb homolog-like, NUMBLIKE, numb-like protein, NUMB-R, Numb-related protein, NUMBRNBL, TNRC23 | |
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human numb (Drosophila) homolog-like using the numbering given in entry NP_004747.1 (GeneID 9253). | |
0.1 mL | |
Cell Biology, Neuroscience, Stem Cells | |
9253 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction