Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUMBL Monoclonal Antibody (OTI8D2), TrueMAB™, OriGene
SDP

Catalog No. 502543769
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3769 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3769 Supplier OriGene Supplier No. CF812202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUMBL Monoclonal antibody specifically detects NUMBL in Human samples. It is validated for Western Blot

Specifications

Antigen NUMBL
Applications Western Blot
Classification Monoclonal
Clone OTI8D2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CIITA
Gene Accession No. P33076
Gene Alias CAG3A; CTG3a; NBL; numb homolog (Drosophila)-like; numb homolog-like; NUMB like, endocytic adaptor protein; Numbl; NUMBLIKE; numb-like; numb-like protein; NUMBR; Numb-R; numb-related protein; TNRC23
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 414-724 of human CIITA produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4261
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.