Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUMBL Mouse anti-Human, Clone: 5F6E1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | NUMBL |
|---|---|
| Clone | 5F6E1 |
| Concentration | 1.48 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6589
![]()
|
Proteintech Group Inc
661551IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.Specifications
| NUMBL | |
| 1.48 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CAG3A, CTG3a, NBL, numb homolog (Drosophila) like, Numb like protein, NUMB R, Numb related protein, NUMBL, NUMBLIKE, NUMBR, TNRC23 | |
| NUMBL | |
| IgG2a | |
| Protein A | |
| Antibody |
| 5F6E1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y6R0 | |
| 9253 | |
| NUMBL Fusion Protein Ag16771 | |
| Primary | |
| -20°C | |
| NUMBL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title