Learn More
Thermo Scientific Nunc Edge 2.0 96-Well Plates
Reduce edge effect and minimize plate evaporation using this uniquely designed moat plate simply by filling the surrounding reservoir with media or sterile water.
Manufacturer: Thermo Scientific 167425
Description
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Specifications
Flat | |
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
0.35cm2/well | |
50 | |
1 | |
Sterile | |
400 μL | |
With Lid | |
8 x 12 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Clear | |
50 | |
Low Flange | |
Individually Wrapped | |
Nunclon Delta | |
Standard | |
96 |
Promotions
-
Thermo Scientific
,
Gibco
,
Invitrogen
Get $1,000 in Life Sciences Products with Purchase
Spend more, get more in Thermo Scientific, Applied Biosystems, Invitrogen, and Gibco life sciences products.: 06-30-2021
-
Thermo Scientific
Get a Case of Cell Culture Products with Purchase
Buy two cases of Thermo Scientific Nunc Cell Culture Products and get a third case at no additional cost.: 01-31-2020
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.