Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Nunc™ IVF Dishes
Click to view available options
Diameter (Metric):
35 mm
54 mm
88 mm
Height (Metric):
11 mm
14 mm
15 mm
Culture Area:
21.5 cm2
56.7 cm2
8.8 cm2
Description
Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Product features for reliable results:
- Facilitate gas exchange with vented lid
- Available in 35mm, 60mm and 90mm sizes
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Specifications
Specifications
| Certifications/Compliance | CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested |
| Description | IVF Dish, 35mm, Lid w/Airvent |
| Culture Area | 8.8 cm2 |
| Material | Polystyrene (USP Class VI) |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| No. per Case | 500 |
| No. per Pack | 10 |
| Height (Metric) | 11 mm |
| Diameter (Metric) | 35 mm |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction