Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Polystyrene Roller Bottles
GREENER_CHOICE

Catalog No. p-4521985
Click to view available options
Cell Growth Area:
1450 cm2
850 cm2
Bottle Surface:
Pleated, no indent
Smooth, no indent
Closure Type:
Easy on/off, Non-vented Cap
Easy on/off, Vented Cap
Packaging:
2/Pk., 20/Cs.
20/Pk., 20/Cs.
20/Pk., 20/Cs. (Double-bagged)

Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.

Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.

Features:

  • Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
  • One-piece seamless design reduces possibility of leaking through a seam.
  • Shallow and deep indents allow for excellent manual and automated handling.
  • Proprietary manufacturing process allows for greater impact resistance
  • Vertical expanded surfaces version facilitates emptying and reduces retention of product

Recommended for:

  • Cell culture expansion
  • Vaccine production
  • Antibody production
TRUSTED_SUSTAINABILITY

Specifications

Bottle Surface Smooth, no indent
Color Clear
For Use With (Application) Cell Culture Expansion, Vaccine Production, Antibody Production
Material Polystyrene
No. per Case 20
No. per Pack 2
Packaging 2/Pk., 20/Cs.
Sterility Sterile
Cell Growth Area 850 cm2
Closure Included Yes
Graduated Yes
Shape Round
Product Type Roller Bottle
Closure Type Easy on/off, Vented Cap
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.