Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
GSA_VAGREENER_CHOICE

Catalog No. 12565491
Change view
Click to view available options
Material:
Aluminum
Non-woven Rayon
PVC-coated Polypropylene
Plastic
Polyester
Polyethylene
Polyolefin
Polypropylene
Vinyl
For Use With (Application):
Breathable, Cell culture
Cell culture, sterile applications
Cell growth, Bacterial growth
Consistent seal
Crystallography and genotyping
General purpose, ELISA
General purpose, sample detection
Low autofluorescence, ELISA
Luminescence, photosensitive samples
PCR
PCR, fluorescence assays, microscopy
PCR, long-term storage
PCR, qPCR, ELISA, Protein crystallography
PCR, storage in 384 well plates
PCR, storage in 96-well plates
Short-term storage, ELISA
Short-term storage, ELISA, minimizes evaporation and cross contamination
Ventilating and storing cell cultures
Certified RNase/DNase-free:
No
Yes
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Application) Certified RNase/DNase-free
12-565-491 Polypropylene PCR, fluorescence assays, microscopy Yes
12-565-398 Aluminum PCR, storage in 96-well plates Yes
12-565-399 Aluminum PCR, storage in 384 well plates Yes
12-565-492 Polypropylene PCR Yes
12-565-512 Polyethylene Low autofluorescence, ELISA No
12-565-513 Polyolefin PCR, qPCR, ELISA, Protein crystallography Yes
12-565-171 Vinyl Luminescence, photosensitive samples No
12-565-71 Polyester Cell culture, sterile applications No
12-567-90 Polyester General purpose, sample detection No
12-566-914 Polyester General purpose, ELISA Yes
12-567-05 Non-woven Rayon Breathable, Cell culture No
12-249-720 Non-woven Rayon Cell growth, Bacterial growth Yes
12-565-463 Plastic Consistent seal No
12-565-475 Aluminum PCR, long-term storage Yes
14-245-18 Polyester Short-term storage, ELISA No
14-245-192B Polyester Short-term storage, ELISA, minimizes evaporation and cross contamination No
AB-0718 Polypropylene Ventilating and storing cell cultures No
AB-0580 PVC-coated Polypropylene Crystallography and genotyping No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 missing translation for 'options'
Catalog No. 12-565-491 Supplier Thermo Scientific™ missing translation for 'supplierNo' 232702
Add to Cart
Add to Cart

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description AMPLIFICATION TAPE
Material Polypropylene
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 100
No. per Pack 100
For Use With (Application) PCR, fluorescence assays, microscopy
Content And Storage General; Long term
Product Type Adhesive Seal
Product Line Nunc
Adhesive Acrylic
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -70°C to +100°C
Well Shape All well shapes
Plate Compatibility All Microplates
Incubation Conditions General
Peelable Yes
Fluorescence Low Autofluorescence
Show More Show Less
Can I boil Nunc Sealing Tape?

Our Nunc Stealing Tape is thermostable from -20 degrees C to 100 degrees C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.