Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils

Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Specifications
| Color | Clear |
| Solvent Or Alcohol Resistant | Life Science Reagents Resistant, DMSO Resistant |
| Description | SEALING TAPE |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Case | 100 |
| No. per Pack | 100 |
| For Use With (Application) | PCR |
| Show More |
Frequently Asked Questions (FAQs)
Our Nunc Stealing Tape is thermostable from -20 degrees C to 100 degrees C.
We recommend using the following Nunc products:
- 96-Well Polypropylene DeepWell Storage Plate, 2 mL (Cat. No. 278743) (sterile growth plate)
- Microplate Lid for 96 microwell plate (Cat. No. 264122) (to cover plate during growth)
- Polyolefin Acrylate Sealing Tape (Cat. No. 232701) (to cover plate during mixing)
- Fritted 96-Well Deep Well Filter Plate (Cat. No. 278011) (for lysate clarification)
- Fritted 96-Well Deep Well Filter Plate with Glass Fiber Filter (Cat. No. 278010) (for DNA binding)
- 96-Well Polypropylene DeepWell Storage Plate, 1.3 mL (Cat. No. 260252) (reception plate)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.