Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps
Click to view available options
For Use With (Equipment):
Barcoded Flasks 178983, 132920
Description
Allow constant airflow and minimize chance of contamination with Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Nunc sterile filter caps are made with HDPE and Thermo Scientific™ GORE™ dPTFE membrane.
- Filter membrane provides effective barrier against microorganisms
- Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
- Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
- Individually wrapped to ensure sterility
Specifications
Specifications
| Closure Color | White |
| Material | HDPE |
| For Use With (Equipment) | Barcoded Flasks 178983, 132920 |
| No. per Case | 100 |
| No. per Pack | 100 |
| Green Features | Fewer resources used and less waste, Sustainable packaging |
| Sterility | Sterile |
| Class | Class VI |
| Closure Type | Filter |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction