Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP107/Nuclear Pore Complex Proteins Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1576 Supplier Bioss Supplier No. BS11678R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-107 is part of the hetero-oligomeric complex that also contains NUP160, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP107 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP107 may be an important drug target in HIV treatment. Multiple isoforms of NUP107 are known to exist.

Specifications

Antigen NUP107/Nuclear Pore Complex Proteins
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NUP107
Gene Accession No. P57740
Gene Alias 107 kDa nucleoporin; AW541137; BcDNA:LD18761; BOS_5256; C76801; CG6743; CG6743 gene product from transcript CG6743-RA; CG6743-PA; Dmel\CG6743; Dmel_CG6743; dmNup107; NPHS11; Nuclear pore complex protein Nup107; nucleoporin 107; nucleoporin 107kD; nucleoporin 107kDa; Nucleoporin Nup107; Nup 107; Nup107; Nup107-PA; Nup170; NUP84; p105
Gene Symbols NUP107
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nuclear pore complex protein Nup107.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57122
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.