Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP107 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598022
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598022 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598022 Supplier Invitrogen™ Supplier No. PA598022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUP107 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-107 is part of the hetero-oligomeric complex that also contains NUP160, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP107 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP107 may be an important drug target in HIV treatment. Multiple isoforms of NUP107 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP107
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NUP107
Gene Accession No. P57740, Q8BH74
Gene Alias 107 kDa nucleoporin; AW541137; BcDNA.LD18761; BcDNA:LD18761; C76801; CG6743; CG6743-PA; Dmel\CG6743; Dmel_CG6743; dmNup107; NPHS11; nuclear pore complex; Nuclear pore complex protein Nup107; Nuclear pore complex protein Nup107-like protein; nucleoporin 107; Nucleoporin 107kD; nucleoporin 107kDa; nucleoporin Nup107; Nup; Nup 107; nup107; Nup107-PA; Nup170; NUP84; p105
Gene Symbols NUP107
Host Species Rabbit
Immunogen Recombinant Human Nuclear pore complex protein Nup107 protein (1-66AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103468, 57122
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.