Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nup133 Rat anti-Mouse, Clone: 3C11G6, MilliporeSigma™
Rat Monoclonal Antibody
$182.31 - $463.74
Specifications
Antigen | Nup133 |
---|---|
Clone | 3C11G6 |
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE192725
|
MilliporeSigma
MABE192725UL |
25 μL |
Each for $182.31
|
|
|||||
MABE1927100
|
MilliporeSigma
MABE1927100UL |
100 μL |
Each for $463.74
|
|
|||||
Description
Nup133 Monoclonal antibody specifically detects Nup133 in Mouse samples. It is validated for Western Blotting.Specifications
Nup133 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG2a κ | |
Protein G purified |
3C11G6 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
His-tagged recombinant fragment corresponding to 447 amino acids from the N-terminal half of murine Nuclear pore complex protein Nup133. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title