Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP153 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555719
Description
Immunogen sequence: GQNREQRESG FSYPNFSLPA ANGLSSGVGG GGGKMRRERT RFVASKPLEE EEMEVPVLPK ISLPITSSSL PTFNFSSPEI TTSSPSPINS SQALTNKVQM TSP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
Nuclear pore complexes regulate the transport of macromolecules between the nucleus and cytoplasm. They are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. Nucleoporins are glycoproteins found in nuclear pores and contain characteristic pentapeptide XFXFG repeats as well as O-linked N-acetylglucosamine residues oriented towards the cytoplasm. The protein encoded by this gene has three distinct domains: a N-terminal region containing a pore targeting and an RNA-binding domain, a central region containing multiple zinc finger motifs, and a C-terminal region containing multiple XFXFG repeats. Alternative splicing results in multiple transcript variants of this gene.
Specifications
NUP153 | |
Polyclonal | |
Unconjugated | |
NUP153 | |
153 kDa nucleoporin; B130015D15Rik; C88147; HNUP153; N153; nuclear pore complex protein hnup153; nuclear pore complex protein Nup153; nucleoporin 153; nucleoporin 153kD; nucleoporin 153kDa; nucleoporin 153kDa L homeolog; nucleoporin Nup153; nucleoporin Nup153 homolog; NUCZINK; nup153; nup153.L; XELAEV_18031694mg | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9972 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49790 | |
NUP153 | |
Recombinant protein corresponding to Human NUP153. Recombinant protein control fragment (Product #RP-94810). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction