Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP153 Recombinant Rabbit Monoclonal Antibody (8D4)
GREENER_CHOICE

Catalog No. FEMA549893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549893 Supplier Invitrogen™ Supplier No. MA549893
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

NUP153 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), ELISA

Nuclear pore complexes regulate the transport of macromolecules between the nucleus and cytoplasm. They are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. Nucleoporins are glycoproteins found in nuclear pores and contain characteristic pentapeptide XFXFG repeats as well as O-linked N-acetylglucosamine residues oriented towards the cytoplasm. The protein encoded by this gene has three distinct domains: a N-terminal region containing a pore targeting and an RNA-binding domain, a central region containing multiple zinc finger motifs, and a C-terminal region containing multiple XFXFG repeats. Alternative splicing results in multiple transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP153
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8D4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NUP153
Gene Accession No. P49790
Gene Alias 153 kDa nucleoporin; B130015D15Rik; C88147; HNUP153; N153; nuclear pore complex protein hnup153; nuclear pore complex protein Nup153; nucleoporin 153; nucleoporin 153kD; nucleoporin 153kDa; nucleoporin 153kDa L homeolog; nucleoporin Nup153; nucleoporin Nup153 homolog; NUCZINK; nup153; nup153.L; XELAEV_18031694mg
Gene Symbols NUP153
Host Species Rabbit
Immunogen A synthesized peptide derived from human Nup153.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9972
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.