Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP155 Mouse anti-Human, Rat, Clone: 1A7F2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NUP155 |
---|---|
Clone | 1A7F2 |
Concentration | 1.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6790
![]() ![]() |
Proteintech Group Inc
663591IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a centrosomal and microtubule-binding protein which is predicted to have two coiled-coil domains and a rhodanese domain. In human retinal pigment epithelial cells the protein localized to centrioles and cilia. Mutations in this gene have been associated with Joubert Syndrome 15; an autosomal recessive ciliopathy and neurological disorder. Alternative splicing results in multiple transcript variants.Specifications
NUP155 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
155 kDa nucleoporin, KIAA0791, N155, nucleoporin 155kDa, Nucleoporin Nup155, NUP155 | |
NUP155 | |
IgG2a | |
Protein A | |
Antibody |
1A7F2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
O75694, P37199 | |
117021, 9631 | |
NUP155 Fusion Protein Ag3638 | |
Primary | |
-20°C | |
NUP155 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title