Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP155 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531786 Supplier Invitrogen™ Supplier No. PA531786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG MCF-7, 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (85%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP155
Applications Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene NUP155
Gene Accession No. O75694
Gene Alias 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140
Gene Symbols NUP155
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human NUP155. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.