Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP155 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596795 Supplier Invitrogen™ Supplier No. PA596795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADMHSTEISL QQRLEYIARA ILSAKSSTAI SSIAADGEFL HELEEKMEVA RIQLQIQETL QRQYSHHSSV QDAVSQLDSE LMDITKLYGE FADPFKLAEC KLAIIHCAGY SDPILVQTLW QDIIEKELSD SVTLSSSDRM HALSLKIVLL GKIYAGTPRF FPLDFIVQFL EQQVCTLNWD VGFVIQTMNE IGVPLPRLLE VYDQLFKSRD PFWNRMKKPL HLLDCIHVLL IRYVENPSQV LNCERRRFTN LCLDAVCGYL VELQSMSSSV AVQAITGNFK SLQAKLERLH; Positive Samples: 293T, HeLa, Jurkat, Mouse liver, Mouse testis; Cellular Location: Cytoplasmic side, Nucleoplasmic side, Nucleus, Nucleus membrane, Peripheral membrane protein, nuclear pore complex.

Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP155
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NUP155
Gene Accession No. O75694
Gene Alias 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140
Gene Symbols NUP155
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1102-1391 of human NUP155 (NP_7056181).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9631
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.