Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP160 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117155 Supplier Invitrogen™ Supplier No. PA5117155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, A-431, Jurkat, HepG2 Immunogen sequence: ALERSFVELS GAERERPRHF REFTVCSIGT ANAVAGAVKY SESAGGFYYV ESGKLFSVTR NRFIHWKTSG DTLELMEESL DINLLNNAIR LKFQNCSVLP GGVYVSETQN R.

The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-160 is part of the hetero-oligomeric complex that also contains NUP107, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP160 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP160 may be an important drug target in HIV treatment. Multiple isoforms of NUP160 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP160
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NUP160
Gene Accession No. Q12769
Gene Alias 160 kDa nucleoporin; 160kDa; 2810011M03Rik; AA414952; AU020188; Gene trap locus 1-13 protein; gene trap locus-13; Gtl1-13; Gtl13; GTL-13; KIAA0197; mKIAA0197; Nuclear pore complex protein Nup160; nucleoporin 160; nucleoporin 160kD; nucleoporin 160kDa; nucleoporin Nup160; NUP120; Nup160
Gene Symbols NUP160
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-150 of human NUP160 (NP_0560461).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23279
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.