Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP188 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128748R
Description
NUP188 Polyclonal antibody specifically detects NUP188 in Human samples. It is validated for Western BlotSpecifications
NUP188 | |
Polyclonal | |
Western Blot | |
FLJ21639, KIAA0169hNup188, nucleoporin 188kDa, nucleoporin NUP188 homolog | |
The immunogen recognized by this antibody maps to a region between residue 1600 and 1650 of human nucleoporin 188kDa using the numbering given in entry NP_056169.1 (GeneID 23511). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23511 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction