Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP188 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566645 Supplier Invitrogen™ Supplier No. PA566645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APMSVYACLGN DAAAIRDAFL TRLQSKIEDM RIKVMILEFL TVAVETQPGL IELFLNLEVK DGSDGSKEFS LGMWSCLHAV LELIDSQQQD R Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

May function as a component of the nuclear pore complex (NPC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP188
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUP188
Gene Accession No. Q5SRE5
Gene Alias BC025526; hNup188; KIAA0169; mKIAA0169; nucleoporin 188; nucleoporin 188kDa; nucleoporin NUP188 homolog; NUP188; U89435
Gene Symbols NUP188
Host Species Rabbit
Immunogen Recombinant Human NUP188. Recombinant protein control fragment (Product #RP-107298).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23511
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.