Learn More
NUP188 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302322AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May function as a component of the nuclear pore complex (NPC).Specifications
NUP188 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5SRE5 | |
NUP188 | |
Between 1600 and 1650 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NUP188 | |
Nucleoporin NUP188 homolog, hNup188, NUP188, KIAA0169 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23511 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.