Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP205 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103979 Supplier Invitrogen™ Supplier No. PA5103979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NUP205.

This gene encodes a nucleoporin, which is a subunit of the nuclear pore complex that functions in active transport of proteins, RNAs and ribonucleoprotein particles between the nucleus and cytoplasm. Mutations in this gene are associated with steroid-resistant nephrotic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP205
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NUP205
Gene Accession No. Q92621
Gene Alias 205 kDa nucleoporin; 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; Nuclear pore complex protein Nup205; nucleoporin 205; nucleoporin 205kDa; nucleoporin Nup205; NUP205
Gene Symbols NUP205
Host Species Rabbit
Immunogen A synthesized peptide derived from human NUP205(Accession Q92621), corresponding to amino acid residues L1129-R1179.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23165, 362335, 70699
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.