Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP205 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116835 Supplier Invitrogen™ Supplier No. PA5116835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, Mouse brain, Rat brain Immunogen sequence: AASLWGPYKD IWHKVGNALW RRQPEAVHLL DKILKKHKPD FISLFKNPPK NVQQHEKVQK ASTEGVAIQG QQGTRLLPEQ LIKEAFILSD LFDIGELAAV ELLLAGEHQQ P.

This gene encodes a nucleoporin, which is a subunit of the nuclear pore complex that functions in active transport of proteins, RNAs and ribonucleoprotein particles between the nucleus and cytoplasm. Mutations in this gene are associated with steroid-resistant nephrotic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP205
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NUP205
Gene Accession No. Q92621
Gene Alias 205 kDa nucleoporin; 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; Nuclear pore complex protein Nup205; nucleoporin 205; nucleoporin 205kDa; nucleoporin Nup205; NUP205
Gene Symbols NUP205
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-120 of human NUP205 (NP_0559501).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23165, 362335, 70699
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.