Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP210 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23907825UL
Description
NUP210 Polyclonal specifically detects NUP210 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NUP210 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q8TEM1 | |
NUP210 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TFTVHFHDNSGDVFHAHSSVLNFATNRDDFVQIGKGPTNNTCVVRTVSVGLTLLRVWDAEHPGLSDFMPLPVLQAISPELSGAMVVGDVLCLATVLTSLEGLSGTWSSSANSILHIDPKTGVAV | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ22389, GP210Pore membrane protein of 210 kDa, KIAA0906Nuclear pore protein gp210, Nuclear envelope pore membrane protein POM 210, nuclear pore membrane glycoprotein 210, nucleoporin 210kDa, POM210Nucleoporin Nup210 | |
Rabbit | |
Affinity Purified | |
RUO | |
23225 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction