Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP214 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111282
Description
Immunogen sequence: FDSPEELPKE RSSLLAVSNK YGLVFAGGAS GLQIFPTKNL LIQNKPGDDP NKIVDKVQGL LVPMKFPIHH LALSCDNLTL SACMMSSEY Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 85%.
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. This gene is a member of the FG-repeat-containing nucleoporins. The protein encoded by this gene is localized to the cytoplasmic face of the nuclear pore complex where it is required for proper cell cycle progression and nucleocytoplasmic transport. The 3' portion of this gene forms a fusion gene with the DEK gene on chromosome 6 in a t(6,9) translocation associated with acute myeloid leukemia and myelodysplastic syndrome. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
| NUP214 | |
| Polyclonal | |
| Unconjugated | |
| NUP214 | |
| 214 kDa nucleoporin; BC039282; CAIN; CAN; CAN protein, putative oncogene; D2H9S46E; Kiaa0023; Nuclear pore complex protein Nup214; nucleoporin 214; nucleoporin 214kDa; nucleoporin Nup214; Nup214; Protein CAN; RGD1304977 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8021 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35658 | |
| NUP214 | |
| Recombinant Protein corresonding to Human NUP214. Recombinant protein control fragment (Product #RP-106203). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction