Learn More
NUP214 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | NUP214 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1494
![]() |
Bethyl Laboratories, Inc
IHC00103T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1493
![]() |
Bethyl Laboratories, Inc
IHC00103 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla and Chimpanzee
Calcineurin is a widely distributed Ser/Thr, calcium and calmodulin-dependent protein phosphatase. It mediates the immunosuppressive actions of drugs such as FK506 and cyclosporin, and has also been implicated in a number of calcium-sensitive pathways in the nervous system. Calcineurin has been found to be associated with other proteins, such as calmodulin, FKBP12, the ryanodine receptor, inositol 1,4,5-trisphosphate, and a recently identified protein, calcineurin inhibitor (cain). Cain has been shown to bind calcineurin through its C-terminal tail. Recent studies have shown cain to bind to both amphiphysin 1 and calcineurin simultaneously and therefore acts as a physiological inhibitor of calcineurin. The binding of cain to amphiphysin 1 does not affect the interaction of amphiphysin 1 with other endocytic proteins. Cain, like calcineurin, is highly expressed in neurons and other tissues.Specifications
NUP214 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Calcineurin Inhibitor | |
NUP214 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P35658 | |
8021 | |
Between 1250 and 1300 | |
Primary | |
4° C | |
NUP214 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.