Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582626 Supplier Invitrogen™ Supplier No. PA582626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAFAVEPQG PALGSEPMML GSPTSPKPGV NAQFLPGFLM GDLPAPVTPQ PRSISGPSVG VMEMRSPLLA GGSPPQ Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 93%.

Nucleoporin Nup35 or Mitotic phosphoprotein 44 is a nuclear protein known to be present on nuclear pore complex and is tightly associated with the nuclear membrane and lamina. This RNA associated protein belongs to the Nup53 family and has a MPPN (mitotic phosphoprotein N' end) domain. It has a small number of scattered phenylalanine-glycine repeats, but not the large domains seen in other nucleoporins. Though the exact function of NUP35 is yet to be revealed, it may function as a component of the nuclear pore complex (NPC) which are collectively referred to as nucleoporins (NUPs). It also may play the role of both NPC structural components and of docking or interaction partners for transiently associated nuclear transport factors, and may also play an important part in the association of MAD1, a transcriptional repressor with the NPC. NUP35 is widely expressed in most of the tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUP35
Gene Accession No. Q8NFH5
Gene Alias 2310006I24Rik; 35 kDa nucleoporin; 35kDa; 5330402E05Rik; Mitotic phosphoprotein 44; Mp44; MP-44; NO44; NP44; Nuclear pore complex protein Nup53; nucleoporin 35; nucleoporin 35kDa; nucleoporin NUP35; Nucleoporin NUP53; Nup35; NUP53
Gene Symbols NUP35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUP35. Recombinant protein control fragment (Product #RP-91854).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129401
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.