Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP35 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571760
Description
Nucleoporin Nup35, or Mitotic phosphoprotein 44, is a nuclear protein known to be present on the nuclear pore complex and is tightly associated with the nuclear membrane and lamina. This RNA-associated protein belongs to the Nup53 family and has a MPPN (mitotic phosphoprotein N′ end) domain. It has a small number of scattered phenylalanine-glycine repeats, but not the large domains seen in other nucleoporins. Though the exact function of NUP35 is yet to be revealed, it may function as a component of the nuclear pore complex (NPC) which are collectively referred to as nucleoporins (NUPs). It also may play the role of both NPC structural components and of docking or interaction partners for transiently associated nuclear transport factors, and may also play an important part in the association of MAD1, a transcriptional repressor with the NPC. NUP35 is widely expressed in most of the tissues.
Specifications
NUP35 | |
Polyclonal | |
Unconjugated | |
NUP35 | |
Nucleoporin NUP53; 35 kDa nucleoporin; Mitotic phosphoprotein 44; MP-44; Nuclear pore complex protein Nup53; Nucleoporin Nup35; NUP35; MP44; NUP53 | |
Rabbit | |
Protein A | |
RUO | |
129401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
1.23 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NFH5 | |
NUP35 | |
KLH conjugated synthetic peptide between 289-318 amino acids from the C-terminal region of human NUP35 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction