Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569404
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%.
Component of the Nup107-160 subcomplex of the nuclear pore complex (NPC). The Nup107-160 subcomplex is required for the assembly of a functional NPC. The Nup107-160 subcomplex is also required for normal kinetochore microtubule attachment, mitotic progression and chromosome segregation.
Specifications
NUP37 | |
Polyclonal | |
Unconjugated | |
NUP37 | |
2410003L22Rik; 2810039M17Rik; nucleoporin 37; nucleoporin 37kDa; nucleoporin Nup37; nucleoporin Nup37-like protein; nup107-160 subcomplex subunit Nup37; NUP37; p37 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
69736 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CWU9 | |
NUP37 | |
synthetic peptide corresponding to a region of mouse NUP37 (aa 53-102). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction