Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP43 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548845
Description
Component of the Nup107-160 subcomplex of the nuclear pore complex (NPC). The Nup107-160 subcomplex is required for the assembly of a functional NPC. The Nup107-160 subcomplex is also required for normal kinetochore microtubule attachment, mitotic progression and chromosome segregation.
Specifications
NUP43 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8NFH3 | |
NUP43 | |
KLH conjugated synthetic peptide between 142-171 amino acids from the Central region of human NUP43. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NUP43 | |
Nucleoporin Nup43, Nup107-160 subcomplex subunit Nup43, p42, NUP43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
348995 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction