Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP43 Monoclonal Antibody (OTI7G10)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526884
Description
NUP43 Monoclonal Antibody for Western Blot, IHC (P)
Component of the Nup107-160 subcomplex of the nuclear pore complex (NPC). The Nup107-160 subcomplex is required for the assembly of a functional NPC. The Nup107-160 subcomplex is also required for normal kinetochore microtubule attachment, mitotic progression and chromosome segregation.
Specifications
| NUP43 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NFH3 | |
| NUP43 | |
| Human recombinant protein fragment corresponding to amino acids 1-253 of NUP43 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7G10 | |
| Unconjugated | |
| NUP43 | |
| 2610016K01Rik; 2610529I12Rik; AA409950; bA350J20.1; nucleoporin 43; nucleoporin 43kDa; nucleoporin Nup43; Nup107-160 subcomplex subunit Nup43; Nup43; p42 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 348995 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction