Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP50 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297442C
Description
NUP50 Polyclonal specifically detects NUP50 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NUP50 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
10762 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50 kDa nucleoporin, MGC39961, NPAP60, NPAP60LNucleoporin Nup50, nuclear pore complex protein Nup50, Nuclear pore-associated protein 60 kDa-like, nuclear pore-associated protein 60L, nucleoporin 50kD, nucleoporin 50kDa | |
Produced in rabbits immunized with E. coli-derived Human NUP50 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction